KLRG1 Monoclonal Antibody (13F12F2), PerCP-eFluor™ 710, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (13F12F2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (13F12F2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9488-42, 46-9488-41)