TIGIT Monoclonal Antibody (MBSA43), PerCP-eFluor™ 710, eBioscience™ - FAQs
TIGIT Monoclonal Antibody (MBSA43), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TIGIT Monoclonal Antibody (MBSA43), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9500-42, 46-9500-41)