TIGIT Monoclonal Antibody (GIGD7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for TIGIT Monoclonal Antibody (GIGD7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9501-82, 46-9501-80)