SEMA4A Monoclonal Antibody (5E3), PerCP-eFluor™ 710, eBioscience™ - FAQs
SEMA4A Monoclonal Antibody (5E3), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for SEMA4A Monoclonal Antibody (5E3), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9753-42, 46-9753-41)