CD303a Monoclonal Antibody (201A), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD303a Monoclonal Antibody (201A), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD303a Monoclonal Antibody (201A), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9818-42, 46-9818-41)