LAP (Latency Associated peptide) Monoclonal Antibody (FNLAP), PerCP-eFluor™ 710, eBioscience™ - FAQs
LAP (Latency Associated peptide) Monoclonal Antibody (FNLAP), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for LAP (Latency Associated peptide) Monoclonal Antibody (FNLAP), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9829-42)