IRF8 Monoclonal Antibody (V3GYWCH), PerCP-eFluor™ 710, eBioscience™ - FAQs
IRF8 Monoclonal Antibody (V3GYWCH), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IRF8 Monoclonal Antibody (V3GYWCH), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9852-82, 46-9852-80)