IRF4 Monoclonal Antibody (3E4), PerCP-eFluor™ 710, eBioscience™ - FAQs
IRF4 Monoclonal Antibody (3E4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IRF4 Monoclonal Antibody (3E4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9858-82, 46-9858-80)