BATF Monoclonal Antibody (MBM7C7), PerCP-eFluor™ 710, eBioscience™ - FAQs
BATF Monoclonal Antibody (MBM7C7), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for BATF Monoclonal Antibody (MBM7C7), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9860-42, 46-9860-41)