IgD Monoclonal Antibody (IA6-2), PerCP-eFluor™ 710, eBioscience™ - FAQs
IgD Monoclonal Antibody (IA6-2), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for IgD Monoclonal Antibody (IA6-2), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9868-41, 46-9868-42)