BCL6 Monoclonal Antibody (BCL-UP), PerCP-eFluor™ 710, eBioscience™ - FAQs
BCL6 Monoclonal Antibody (BCL-UP), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for BCL6 Monoclonal Antibody (BCL-UP), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9880-42, 46-9880-41)