GARP Monoclonal Antibody (G14D9), PerCP-eFluor™ 710, eBioscience™ - FAQs
GARP Monoclonal Antibody (G14D9), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for GARP Monoclonal Antibody (G14D9), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9882-41, 46-9882-42)