HELIOS Monoclonal Antibody (22F6), PerCP-eFluor™ 710, eBioscience™ - FAQs
HELIOS Monoclonal Antibody (22F6), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for HELIOS Monoclonal Antibody (22F6), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9883-42, 46-9883-41)