GARP Monoclonal Antibody (YGIC86), PerCP-eFluor™ 710, eBioscience™ - FAQs
GARP Monoclonal Antibody (YGIC86), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for GARP Monoclonal Antibody (YGIC86), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9891-82, 46-9891-80)