CD278 (ICOS) Monoclonal Antibody (15F9), PerCP-eFluor 710, eBioscience™ - FAQs
CD278 (ICOS) Monoclonal Antibody (15F9), PerCP-eFluor 710, eBioscience™ - FAQs
View additional product information for CD278 (ICOS) Monoclonal Antibody (15F9), PerCP-eFluor 710, eBioscience™ - FAQs (46-9940-80)