HLA-DR Monoclonal Antibody (L243), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for HLA-DR Monoclonal Antibody (L243), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9952-41, 46-9952-42)