HLA-G Monoclonal Antibody (87G), PerCP-eFluor™ 710, eBioscience™ - FAQs
HLA-G Monoclonal Antibody (87G), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for HLA-G Monoclonal Antibody (87G), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9957-42, 46-9957-41)