HLA-ABC Monoclonal Antibody (W6/32), PerCP-eFluor™ 710, eBioscience™ - FAQs
HLA-ABC Monoclonal Antibody (W6/32), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for HLA-ABC Monoclonal Antibody (W6/32), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9983-42)