CD279 (PD-1) Monoclonal Antibody (J43), PerCP-eFluor™ 710, eBioscience™ - FAQs
CD279 (PD-1) Monoclonal Antibody (J43), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (J43), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9985-80, 46-9985-82)