Human IgM Monoclonal Antibody (SA-DA4), PerCP-eFluor™ 710, eBioscience™ - FAQs
Human IgM Monoclonal Antibody (SA-DA4), PerCP-eFluor™ 710, eBioscience™ - FAQs
View additional product information for Human IgM Monoclonal Antibody (SA-DA4), PerCP-eFluor™ 710, eBioscience™ - FAQs (46-9998-41, 46-9998-42)