Succinylcholine chloride, 96%
Succinylcholine chloride, 96%
Succinylcholine chloride, 96%
Thermo Scientific Chemicals

Succinylcholine chloride, 96%

CAS: 71-27-2 | C14H30Cl2N2O4 | 361.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 460111000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS71-27-2
IUPAC Nametrimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dichloride
Molecular FormulaC14H30Cl2N2O4
InChI KeyYOEWQQVKRJEPAE-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point160°C to 166°C
Titration Argentometric>=95 % (on dry substance)
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only