4-Chloro-1-butene, 98%, Thermo Scientific Chemicals
4-Chloro-1-butene, 98%, Thermo Scientific Chemicals
4-Chloro-1-butene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-1-butene, 98%, Thermo Scientific Chemicals

CAS: 927-73-1 | C4H7Cl | 90.55 g/mol
Have Questions?
Catalog NumberQuantity
4601600505 g
Catalog number 460160050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS927-73-1
IUPAC Name4-chlorobut-1-ene
Molecular FormulaC4H7Cl
InChI KeyWKEVRZCQFQDCIR-UHFFFAOYSA-N
SMILESClCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=97.5 %
Refractive index1.4220 to 1.4240 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only