Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Frames and Caps for Immuno Standard Modules
Thermo Scientific™

Frames and Caps for Immuno Standard Modules

Easily insert and remove strip plates with solid modules by utilizing these frames and caps for immuno standard modules.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorSterilityProduct Type
460348WhiteNon-sterileFrame
430082ClearNon-sterileStrip Cap
430805ClearSterileStrip Cap
Catalog number 460348
Price (USD)
277.70
Online exclusive
364.39
Save 86.69
Case of 60
Color:
White
Sterility:
Non-sterile
Product Type:
Frame
Price (USD)
277.70
Online exclusive
364.39
Save 86.69
Case of 60

Easily insert and remove strip plates with solid modules by utilizing Thermo Scientific™ Frames and Caps for Immuno Standard Modules.

  • Polyethylene, stackable frame is marked for lid orientation
Specifications
For Use With (Equipment)Nunc Modules
No. per Case60
No. per Pack5
Plate CompatibilityImmuno Standard Modules
Product TypeFrame
Quantity60/Cs.
ColorWhite
MaterialPolyethylene
SterilityNon-sterile
Unit SizeCase of 60