White and Black 384-Well Immuno Plates
White and Black 384-Well Immuno Plates
White and Black 384-Well Immuno Plates
White and Black 384-Well Immuno Plates
Thermo Scientific™

White and Black 384-Well Immuno Plates

Conduct quantitative and qualitative immunoassays with these white and black 384-well immuno plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
460372White
460518Black
Catalog number 460372
Price (USD)
285.93
Online exclusive
333.94
Save 48.01
Case of 30
Color:
White
Price (USD)
285.93
Online exclusive
333.94
Save 48.01
Case of 30

Conduct quantitative and qualitative immunoassays with Thermo Scientific™ White and Black 384-Well Immuno Plates. Ideal for ELISA and binding assays, these plates are compatible with common instruments. They offer lot-to-lot reliability and well-to-well reproducibility for optimal results.

Features:

  • Plate has SBS footprint, fits standard equipment and is available with flat-bottom wells
  • Thermo Scientific MaxiSorp is hydrophilic and ideal for antibody sandwich assays
  • Plate is offered as black or white for optimal choice of application type
  • Certified binding offers homogeneity/reproducibility.

White:

  • Maximum reflection/minimum autofluorescence and autoluminescence

Black:

  • Minimizes crosstalk, back-scattered light and background in fluorescence reading

Compatible with:

Lids for Thermo Scientific MicroWell plates, sealing tape and breathable membrane

Ordering information:

Availability may vary by country

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Binding PropertyHydrophilic
Detection MethodChemiluminescence
For Use With (Application)Fluorescence and Luminescence
FormatStandard
LidWithout Lid
No. per Case30
No. per Pack10
Packaging30 Cases
Plate BlockingNone
Product TypeMicroplate
Target MoleculeAntibodies (Glycoproteins)
Volume (Metric) Working100 μL
Well DesignF384
Array16 x 24
ColorWhite
MaterialPolystyrene
No. of Wells384
SterilityNon-sterile
Surface TreatmentMaxiSorp
Well ShapeSquare, Flat Bottom (F-well)
Unit SizeCase of 30

Frequently asked questions (FAQs)

For Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, is the surface treatment solely on the bottom of the well, or is it also applied to the sides?

For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.

What length of peptide is ideal for binding to the Thermo Scientific MaxiSorp surface? What are the detection limitations?

We have tested and found that a 3 amino acid peptide (Pro, Leu, Gly) cannot be detected when passively adsorbed on the MaxiSorp surface. However, this peptide can be detected when covalently immobilized using CovaLink NH Modules and CovaLink NH2 Modules and Plates. Using covalent immobilization of small peptide residues, you can typically obtain a better orientation of the molecule and reduced problems with antibody recognition of the peptide due to masking of the epitope. We have discovered that a 7 amino acid peptide from the MHC Class II antigen can be detected when adsorbed on the MaxiSorp surface. We state that the detection limitation using the MaxiSorp surface is between 3 and 7 amino acid residues.
One additional note is that detection is contingent upon the orientation of the peptide when immobilized. If the active site is inactivated or hidden at the site facing the solid phase, no detection signal is observed.

Can I bind either single- or double-stranded DNA to the Thermo Scientific MaxiSorp surface?

Single-stranded DNA can be adsorbed to our MaxiSorp surface using approximately 10 µg ssDNA per mL PBS, pH 8.2, although the stability is uncertain. Based on our experience, ssDNA immobilized on the MaxiSorp surface is so loosely bound that it can be removed by stringent washing.
Double-stranded DNA will not bind to the MaxiSorp surface. DNA, however, can be covalently bound to Nunc NucleoLink Strips.

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.

For Nunc Immuno plates and modules, what are the advantages of one well geometry type over another? Which types should I use for various applications?

The following list describes the geometries of wells available for Nunc Immuno-plates and modules:
- Flat bottom (F): Allows maximum transmission of light. These plates can be read on a monochromatic reader.
- Round bottom (U): This geometry optimizes washing and coating. These plates can be read using a dual wavelength reader.
- "C" bottom (C): This design of the well is a combination of both flat and round bottoms. Basically, it is a flat bottomed well with curved edges at the bottom. These plates also can be read using a monochromatic reader combining optimal reading and washing.
- StarWell: These wells have a modified "C" shape geometry with eight fins strategically placed at the bottom. This feature increases surface area, allowing more molecules to become immobilized which reduces incubation times.