Daidzin, 98%
Daidzin, 98%
Daidzin, 98%
Thermo Scientific Chemicals

Daidzin, 98%

CAS: 552-66-9 | C21H20O9 | 416.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number 460420010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS552-66-9
IUPAC Name3-(4-hydroxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4H-chromen-4-one
Molecular FormulaC21H20O9
InChI KeyKYQZWONCHDNPDP-QNDFHXLGSA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=C3C(=O)C(=COC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)White to light yellow
Loss on drying=<2.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only