2-Bromobornane, technical
2-Bromobornane, technical
2-Bromobornane, technical
Thermo Scientific Chemicals

2-Bromobornane, technical

CAS: 4443-48-5 | C10H17Br | 217.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 460740250
Price (USD)
183.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
183.00
Each
Chemical Identifiers
CAS4443-48-5
IUPAC Name2-bromo-1,7,7-trimethylbicyclo[2.2.1]heptane
Molecular FormulaC10H17Br
InChI KeyOTOQMOVZIUGCQE-UHFFFAOYNA-N
SMILESCC1(C)C2CCC1(C)C(Br)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=78.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only