Azaperone
Azaperone
Azaperone
Thermo Scientific Chemicals

Azaperone

CAS: 1649-18-9 | C19H22FN3O | 327.40 g/mol
Have Questions?
Catalog NumberQuantity
4608300101 g
Catalog number 460830010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS1649-18-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Loss on drying=<0.5 % (60°C, 4 h) (vacuum)
Melting point92°C to 95°C
Titration with HClO499 to 101 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only