Anastrozole, 98%
Anastrozole, 98%
Anastrozole, 98%
Thermo Scientific Chemicals

Anastrozole, 98%

CAS: 120511-73-1 | C17H19N5 | 293.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 460972500
Price (USD)
217.00
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
217.00
Each
Chemical Identifiers
CAS120511-73-1
IUPAC Name2-[3-(1-cyano-1-methylethyl)-5-[(1H-1,2,4-triazol-1-yl)methyl]phenyl]-2-methylpropanenitrile
Molecular FormulaC17H19N5
InChI KeyYBBLVLTVTVSKRW-UHFFFAOYSA-N
SMILESCC(C)(C#N)C1=CC(=CC(CN2C=NC=N2)=C1)C(C)(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Infrared spectrumConforms
Melting point82°C to 86°C
HPLC>=97.5 %
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only