Carbidopa monohydrate, 99%
Carbidopa monohydrate, 99%
Carbidopa monohydrate, 99%
Thermo Scientific Chemicals

Carbidopa monohydrate, 99%

CAS: 38821-49-7 | C10H14N2O4·H2O | 244.25 g/mol
Have Questions?
Catalog NumberQuantity
4610100101 g
Catalog number 461010010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS38821-49-7
IUPAC Name(2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid hydrate
Molecular FormulaC10H16N2O5
InChI KeyQTAOMKOIBXZKND-PPHPATTJSA-N
SMILESO.C[C@@](CC1=CC=C(O)C(O)=C1)(NN)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying6.9 % to 7.9 % (1 g, 105°C)
HPLC>=98.5 %
Appearance (Form)Powder
Appearance (Color)White to yellowish-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only