Nevirapine, 98%
Nevirapine, 98%
Nevirapine, 98%
Thermo Scientific Chemicals

Nevirapine, 98%

CAS: 129618-40-2 | C15H14N4O | 266.3 g/mol
Have Questions?
Catalog NumberQuantity
461152500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461152500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS129618-40-2
IUPAC Name2-cyclopropyl-7-methyl-2,4,9,15-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3(8),4,6,11,13-hexaen-10-one
Molecular FormulaC15H14N4O
InChI KeyNQDJXKOVJZTUJA-UHFFFAOYSA-N
SMILESCC1=CC=NC2=C1NC(=O)C1=CC=CN=C1N2C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point244°C to 248°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only