Ondansetron hydrochloride dihydrate, 98%
Ondansetron hydrochloride dihydrate, 98%
Ondansetron hydrochloride dihydrate, 98%
Thermo Scientific Chemicals

Ondansetron hydrochloride dihydrate, 98%

CAS: 103639-04-9 | HCl·2H2O | 365.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 461160010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS103639-04-9
IUPAC Namehydrogen 9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-2,3,4,9-tetrahydro-1H-carbazol-4-one dihydrate chloride
Molecular FormulaC18H24ClN3O3
InChI KeyVRSLTNZJOUZKLX-UHFFFAOYNA-N
SMILES[H+].O.O.[Cl-].CN1C2=C(C3=CC=CC=C13)C(=O)C(CN1C=CN=C1C)CC2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point176°C to 180°C
Water=<10.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only