Pyrimethamine, 98%, Thermo Scientific Chemicals
Pyrimethamine, 98%, Thermo Scientific Chemicals
Pyrimethamine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pyrimethamine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4612000101g
4612000505g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461200010
Price (USD)
38.40
Each
Quantity:
1g
Request bulk or custom format
Price (USD)
38.40
Each
Chemical Identifiers
CAS58-14-0
IUPAC Name5-(4-chlorophenyl)-6-ethylpyrimidine-2,4-diamine
Molecular FormulaC12H13ClN4
InChI KeyWKSAUQYGYAYLPV-UHFFFAOYSA-N
SMILESCCC1=NC(N)=NC(N)=C1C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point239°C to 243°C
Loss on drying=<0.5 % (105°C, 4 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only