Product Image
Thermo Scientific Chemicals

Cedrol

222.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 461421000
Price (USD)
251.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
251.00
Each
Chemical Identifiers
CAS77-53-2
IUPAC Name(1S,2R,5S,7R,8R)-2,6,6,8-tetramethyltricyclo[5.3.1.0¹,⁵]undecan-8-ol
Molecular FormulaC15H26O
InChI KeySVURIXNDRWRAFU-OGMFBOKVSA-N
SMILESC[C@@H]1CC[C@H]2C(C)(C)[C@H]3C[C@@]12CC[C@@]3(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystals
Melting point80°C to 86°C
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only