Brimonidine, 98%
Brimonidine, 98%
Brimonidine, 98%
Thermo Scientific Chemicals

Brimonidine, 98%

CAS: 59803-98-4 | C11H10BrN5 | 292.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 461522500
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
102.65
Special offer
Online exclusive
Ends: 28-Dec-2026
120.00
Save 17.35
Each
Chemical Identifiers
CAS59803-98-4
IUPAC Name5-bromo-N-(4,5-dihydro-1H-imidazol-2-yl)quinoxalin-6-amine
Molecular FormulaC11H10BrN5
InChI KeyXYLJNLCSTIOKRM-UHFFFAOYSA-N
SMILESBrC1=C2N=CC=NC2=CC=C1NC1=NCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Appearance (Form)Powder
Infrared spectrumConforms
Residue after ignition=<0.3 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only