Product Image
Thermo Scientific Chemicals

Febuxostat, 98%

CAS: 144060-53-7 | C16H16N2O3S | 316.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 461580010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS144060-53-7
IUPAC Name2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
Molecular FormulaC16H16N2O3S
InChI KeyBQSJTQLCZDPROO-UHFFFAOYSA-N
SMILESCC(C)COC1=CC=C(C=C1C#N)C1=NC(C)=C(S1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
HPLC>=97.5 %
Appearance (Form)Powder
Infrared spectrumConforms
Melting point206°C to 216°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only