Rivaroxaban, 98%
Rivaroxaban, 98%
Rivaroxaban, 98%
Thermo Scientific Chemicals

Rivaroxaban, 98%

CAS: 366789-02-8 | C19H18ClN3O5S | 435.88 g/mol
Have Questions?
Catalog NumberQuantity
4616000101 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461600010
Price (USD)
311.65
Special offer
Online exclusive
Ends: 28-Dec-2026
366.00
Save 54.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
311.65
Special offer
Online exclusive
Ends: 28-Dec-2026
366.00
Save 54.35
Each
Chemical Identifiers
CAS366789-02-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point228°C to 232°C
Loss on drying=<0.5 %
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only