This product ships via ground from US northeast.
Ganciclovir, 98%, Thermo Scientific Chemicals
Ganciclovir, 98%, Thermo Scientific Chemicals
Ganciclovir, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ganciclovir, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4617100101g
4617100505g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461710010
Price (USD)
44.80
Each
Quantity:
1g
Request bulk or custom format
Price (USD)
44.80
Each
Chemical Identifiers
CAS82410-32-0
IUPAC Name2-amino-9-{[(1,3-dihydroxypropan-2-yl)oxy]methyl}-6,9-dihydro-3H-purin-6-one
Molecular FormulaC9H13N5O4
InChI KeyIRSCQMHQWWYFCW-UHFFFAOYSA-N
SMILESNC1=NC(=O)C2=C(N1)N(COC(CO)CO)C=N2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<4 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only