Fenofibrate, 98%
Fenofibrate, 98%
Fenofibrate, 98%
Thermo Scientific Chemicals

Fenofibrate, 98%

CAS: 49562-28-9 | C20H21ClO4 | 360.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 461780250
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS49562-28-9
IUPAC Namepropan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
Molecular FormulaC20H21ClO4
InChI KeyYMTINGFKWWXKFG-UHFFFAOYSA-N
SMILESCC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Melting point79°C to 83°C
Infrared spectrumConforms
Loss on drying=<0.5 % (60°C) (vacuum)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only