Product Image
Thermo Scientific Chemicals

Vinorelbine tartrate, 98%

CAS: 125317-39-7 | C53H66N4O20 | 1079.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
Catalog number 461981000
Price (USD)
530.65
Online exclusive
589.00
Save 58.35
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
530.65
Online exclusive
589.00
Save 58.35
Each
Chemical Identifiers
CAS125317-39-7
IUPAC Namebis((2R,3R)-2,3-dihydroxybutanedioic acid); methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(12S)-16-ethyl-12-(methoxycarbonyl)-1,10-diazatetracyclo[12.3.1.0³,¹¹.0⁴,⁹]octadeca-3(11),4,6,8,15-pentaen-12-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate
Molecular FormulaC53H66N4O20
InChI KeyCILBMBUYJCWATM-NPJYPKOYSA-N
SMILESO[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CCC1=CC2CN(C1)CC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only