Product Image
Thermo Scientific Chemicals

1,2-Bis(trichlorosilyl)ethane, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 462030050
Price (USD)
85.80
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
85.80
Each
Chemical Identifiers
CAS2504-64-5
IUPAC Nametrichloro[2-(trichlorosilyl)ethyl]silane
Molecular FormulaC2H4Cl6Si2
InChI KeyWDVUXWDZTPZIIE-UHFFFAOYSA-N
SMILESCl[Si](Cl)(Cl)CC[Si](Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to light yellow
Appearance (Form)Crystalline mass or liquid
Infrared spectrumConforms
Refractive index1.4735 to 1.4760 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only