Isoprenaline hemisulfate
Isoprenaline hemisulfate
Isoprenaline hemisulfate
Thermo Scientific Chemicals

Isoprenaline hemisulfate

CAS: 299-95-6 | C11H17NO3·1/2H2O4S | 520.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 462110050
Price (USD)
56.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
56.00
Each
Chemical Identifiers
CAS299-95-6
IUPAC Namebis(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}benzene-1,2-diol); sulfuric acid
Molecular FormulaC22H36N2O10S
InChI KeyZOLBALGTFCCTJF-UHFFFAOYNA-N
SMILESOS(O)(=O)=O.CC(C)NCC(O)C1=CC=C(O)C(O)=C1.CC(C)NCC(O)C1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<7 %
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Identitypasses test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only