Fluvoxamine maleate
Thermo Scientific Chemicals

Fluvoxamine maleate

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 462350010
Price (USD)
64.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
64.40
Each
Chemical Identifiers
CAS61718-82-9
IUPAC Name(2Z)-but-2-enedioic acid; (E)-(2-aminoethoxy)({5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene})amine
Molecular FormulaC19H25F3N2O6
InChI KeyLFMYNZPAVPMEGP-PIDGMYBPSA-N
SMILESOC(=O)\C=C/C(O)=O.COCCCC\C(=N/OCCN)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.1 %
Titration with HClO497.5 to 102.5 %
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only