Product Image
Thermo Scientific Chemicals

Sunitinib malate

CAS: 341031-54-7 | C26H33FN4O7 | 532.57 g/mol
Have Questions?
Catalog NumberQuantity
4626400101 g
Catalog number 462640010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS341031-54-7
IUPAC Name(2S)-2-hydroxybutanedioic acid; N-[2-(diethylamino)ethyl]-5-{[(3Z)-5-fluoro-2-oxo-2,3-dihydro-1H-indol-3-ylidene]methyl}-2,4-dimethyl-1H-pyrrole-3-carboxamide
Molecular FormulaC26H33FN4O7
InChI KeyLBWFXVZLPYTWQI-IPOVEDGCSA-N
SMILESO[C@@H](CC(O)=O)C(O)=O.CCN(CC)CCNC(=O)C1=C(C)NC(C=C2/C(=O)NC3=CC=C(F)C=C23)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=98.5 %
Appearance (Color)Yellow to orange
Solubility(1 % in DMSO) Clear yellow to yellow-orange solution
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only