Product Image
Thermo Scientific Chemicals

Eflornithine hydrochloride monohydrate

CAS: 96020-91-6 | C6H15ClF2N2O3 | 236.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number 462700010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS96020-91-6
IUPAC Namehydrogen 2,5-diamino-2-(difluoromethyl)pentanoic acid hydrate chloride
Molecular FormulaC6H15ClF2N2O3
InChI KeyFJPAMFNRCFEGSD-UHFFFAOYNA-N
SMILES[H+].O.[Cl-].NCCCC(N)(C(F)F)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Water6 to 9 %
Titration with HClO4>=97.5 % (dry substance)
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only