Product Image
Thermo Scientific Chemicals

Risedronic acid sodium salt, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 462930010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS115436-72-1
IUPAC Namesodium hydrogen [1-hydroxy-1-phosphono-2-(pyridin-3-yl)ethyl]phosphonate
Molecular FormulaC7H10NNaO7P2
InChI KeyDRFDPXKCEWYIAW-UHFFFAOYNA-M
SMILES[Na+].OC(CC1=CN=CC=C1)(P(O)(O)=O)P(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water11.9 to 13.9 %
HPLC>=97.5 %
Appearance (Color)White to beige
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only