Product Image
Thermo Scientific Chemicals

Silibinin

CAS: 22888-70-6 | C25H22O10 | 482.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 463030050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS22888-70-6
IUPAC Name3,5,7-trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC25H22O10
InChI KeySEBFKMXJBCUCAI-UHFFFAOYNA-N
SMILESCOC1=CC(=CC=C1O)C1OC2=CC(=CC=C2OC1CO)C1OC2=CC(O)=CC(O)=C2C(=O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 % (mixture of A and B)
Loss on drying=<4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only