Product Image
Thermo Scientific Chemicals

Tigecycline

Have Questions?
Catalog NumberQuantity
463062500250 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463062500
Price (USD)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS220620-09-7
IUPAC NameN-[(5aR,6aS,7S,9Z,10aS)-9-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-1,10a,12-trihydroxy-8,10,11-trioxo-5,5a,6,6a,7,8,9,10,10a,11-decahydrotetracen-2-yl]-2-(tert-butylamino)acetamide
Molecular FormulaC29H39N5O8
InChI KeyZXGBRIBPJBHLMO-SBMFAFPZSA-N
SMILESCN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=C(NC(=O)CNC(C)(C)C)C(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to yellow-brown
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Water=<4.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only