Product Image
Thermo Scientific Chemicals

Amifostine trihydrate

CAS: 112901-68-5 | C5H21N2O6PS | 268.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463100010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS112901-68-5
IUPAC Name({2-[(3-aminopropyl)amino]ethyl}sulfanyl)phosphonic acid trihydrate
Molecular FormulaC5H21N2O6PS
InChI KeyTXQPXJKRNHJWAX-UHFFFAOYSA-N
SMILESO.O.O.NCCCNCCSP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Appearance (Color)White to almost white
Water18 to 22 %
Appearance (Form)Powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only