Product Image
Thermo Scientific Chemicals

Duloxetine hydrochloride

333.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 463140050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS136434-34-9
IUPAC Namehydrogen methyl[(3S)-3-(naphthalen-1-yloxy)-3-(thiophen-2-yl)propyl]amine chloride
Molecular FormulaC18H20ClNOS
InChI KeyBFFSMCNJSOPUAY-LMOVPXPDSA-N
SMILES[H+].[Cl-].CNCC[C@H](OC1=C2C=CC=CC2=CC=C1)C1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Melting point162°C to 168°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only