Product Image
Thermo Scientific Chemicals

Sildenafil

CAS: 139755-83-2 | C22H30N6O4S | 474.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 463240010
Price (USD)
293.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
293.00
Each
Chemical Identifiers
CAS139755-83-2
IUPAC Name5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular FormulaC22H30N6O4S
InChI KeyBNRNXUUZRGQAQC-UHFFFAOYSA-N
SMILESCCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<2.5 %
HPLC>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to almost white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only